AOB640091223 AOBChem

4-Benzyloxy-5-bromo-2-(piperidin-1-yl)-pyrimidine

  • MDL number MFCD06798239
  • Cas Number : 885952-22-7
  • Molecular Weight : 348.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/4-benzyloxy-5-bromo-2-piperidin-1-yl-pyrimidine.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/10135.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart