AOB640091437 AOBChem

1-Bromo-4-isopropylbenzene

  • MDL number MFCD00039159
  • Cas Number : 586-61-8
  • Molecular Weight : 199.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/1-bromo-4-isopropylbenzene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/10400.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart