AOB640091572 AOBChem

Isopentylboronic acid

  • MDL number MFCD07784365
  • Cas Number : 98139-72-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96.00
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/isopentylboronic-acid.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/10574.pdf
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart