AOB640091706 AOBChem

4-Butoxy-3-fluorophenylboronic acid

  • MDL number MFCD06411286
  • Cas Number : 156487-13-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/4-butoxy-3-fluorophenylboronic-acid.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/11109.pdf
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart