AOB640091801 AOBChem

5-Chloro-2-methylthiopyridine

  • MDL number MFCD03094947
  • Cas Number : 89379-91-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/5-chloro-2-methylthiopyridine.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/11396.pdf
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart