AOB640092219 AOBChem

2-Chloro-3-cyanopyridine-4-boronic acid

  • MDL number MFCD09037478
  • Cas Number : 878194-88-8
  • Molecular Weight : 182.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/2-chloro-3-cyanopyridine-4-boronic-acid.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/13561.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart