AOB640092246 AOBChem

4-Bromo-1-isopropyl-1H-pyrazole

  • MDL number MFCD09801021
  • Cas Number : 313735-62-5
  • Molecular Weight : 189.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/4-bromo-1-isopropyl-1h-pyrazole.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/13798.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart