AOB640092396 AOBChem

2-Chloro-3-fluoro-5-iodopyridine

  • MDL number MFCD09909856
  • Cas Number : 153034-99-2
  • Molecular Weight : 257.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/2-chloro-3-fluoro-5-iodopyridine.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/14213.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart