AOB640092405 AOBChem

3-Benzyloxy-4-methylbenzoic acid

  • MDL number MFCD06656132
  • Cas Number : 165662-68-0
  • Molecular Weight : 242.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/3-benzyloxy-4-methylbenzoic-acid.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/14253.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart