AOB640092680 AOBChem

1-Bromo-5-chloro-3-fluoro-2-iodobenzene

  • MDL number MFCD00142591
  • Cas Number : 201849-16-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/1-bromo-5-chloro-3-fluoro-2-iodobenzene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/15228.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart