AOB640092891 AOBChem

3-Bromo-5-trifluoromethylbenzaldehyde

  • MDL number MFCD08059506
  • Cas Number : 477535-41-4
  • Molecular Weight : 253.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/3-bromo-5-trifluoromethylbenzaldehyde.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/15865.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart