AOB640092986 AOBChem

2-Bromo-6-fluoro-3-methoxybenzaldehyde

  • MDL number MFCD11856027
  • Cas Number : 154650-22-3
  • Molecular Weight : 233.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/2-bromo-6-fluoro-3-methoxybenzaldehyde.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/16297.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart