AOB640093233 AOBChem

1-Bromo-4-iodo-2-(methylthio)benzene

  • MDL number MFCD11617269
  • Cas Number : 1032231-26-7
  • Molecular Weight : 329

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/1-bromo-4-iodo-2-methylthio-benzene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/17062.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart