AOB640093267 AOBChem

4-(Benzyloxy)-1-bromo-2-fluorobenzene

  • MDL number MFCD09801041
  • Cas Number : 185346-79-6
  • Molecular Weight : 281.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/4-benzyloxy-1-bromo-2-fluorobenzene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/17220.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart