AOB640093313 AOBChem

3-Fluoro-5-(piperidine-1-carbonyl)phenylboronic acid

  • MDL number MFCD08235060
  • Cas Number : 874219-43-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/3-fluoro-5-piperidine-1-carbonyl-phenylboronic-acid.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/17334.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart