AOB640093391 AOBChem

1-Bromo-3-chloro-5-methoxybenzene

  • MDL number MFCD11035921
  • Cas Number : 174913-12-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/1-bromo-3-chloro-5-methoxybenzene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/17597.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart