AOB640093435 AOBChem

2-fluoro-4-methoxy-1-(phenylmethoxy)-benzene

  • MDL number MFCD26045239
  • Cas Number : 540463-60-3
  • Molecular Weight : 232.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/2-fluoro-4-methoxy-1-phenylmethoxy-benzene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/17734.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart