AOB640093454 AOBChem

3-Chloro-N,N-diethylbenzamide

  • MDL number MFCD01214024
  • Cas Number : 15952-65-5
  • Molecular Weight : 211.69

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/3-chloro-n-n-diethylbenzamide.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/17784.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart