AOB640093627 AOBChem

6-Chloro-2-fluoro-3-methoxybenzyl alcohol

  • MDL number MFCD04115949
  • Cas Number : 886499-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/6-chloro-2-fluoro-3-methoxybenzyl-alcohol.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/18379.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart