AOB640093662 AOBChem

5-Chloro-6-methoxypyridine-3-carboxylic acid

  • MDL number MFCD04972405
  • Cas Number : 884494-85-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/5-chloro-6-methoxypyridine-3-carboxylic-acid.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/18477.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart