AOB640093968 AOBChem

2,6-Difluoro-3-nitrophenylboronic acid pinacol ester

  • MDL number MFCD16295275
  • Cas Number : 1451391-10-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/2-6-difluoro-3-nitrophenylboronic-acid-pinacol-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/19645.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart