AOB640093981 AOBChem

4-Bromo-2-iodobenzonitrile

  • MDL number MFCD09907887
  • Cas Number : 944276-67-9
  • Molecular Weight : 307.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/4-bromo-2-iodobenzonitrile.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/19700.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart