AOB640094669 AOBChem

1-Boc-4-fluoro-1H-indazole

  • MDL number MFCD18911747
  • Cas Number : 1305320-65-3
  • Molecular Weight : 236.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/1-boc-4-fluoro-1h-indazole.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/22187.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart