AOB640094911 AOBChem

3-Bromo-4-chlorophenol

  • MDL number MFCD00070740
  • Cas Number : 13659-24-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98.00
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/3-bromo-4-chlorophenol.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/23143.pdf
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart