AOB640094930 AOBChem

1-(4-Bromo-2-chlorophenyl)methyl@piperidine

  • MDL number MFCD22543656
  • Cas Number : 1200131-41-4
  • Molecular Weight : 288.61

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/1-4-bromo-2-chlorophenyl-methylpiperidine.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/23220.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart