AOB640094948 AOBChem

5-Bromo-2-ethoxy-1-fluoro-3-(trifluoromethyl)benzene

  • MDL number MFCD22056755
  • Cas Number : 1370025-61-8
  • Molecular Weight : 287.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/5-bromo-2-ethoxy-1-fluoro-3-trifluoromethyl-benzene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/23313.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart