AOB640095047 AOBChem

1-Fluoro-4-methyl-2-(methylsulfanyl)benzene

  • MDL number MFCD15527259
  • Cas Number : 1370025-55-0
  • Molecular Weight : 156.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/1-fluoro-4-methyl-2-methylsulfanyl-benzene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/23600.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart