AOB640095295 AOBChem

1-Bromo-2-iodo-3,5-dimethylbenzene

  • MDL number MFCD07780646
  • Cas Number : 933671-83-1
  • Molecular Weight : 310.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/1-bromo-2-iodo-3-5-dimethylbenzene.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/24396.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart