AOB640095371 AOBChem

3-Bromo-4-methoxybenzyl alcohol

  • MDL number MFCD01861398
  • Cas Number : 38493-59-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/3-bromo-4-methoxybenzyl-alcohol.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/24533.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart