AOB640095378 AOBChem

4-(N,O-Dimethylhydroxylaminocarbonyl)-2-chlorophenylboronic acid

  • MDL number MFCD22543722
  • Cas Number : 1451392-33-8
  • Molecular Weight : 243.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/4-n-o-dimethylhydroxylaminocarbonyl-2-chlorophenylboronic-acid.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/24544.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart