AOB640095396 AOBChem

5-Bromo-4-methyl-2-(methylthio)pyridine

  • MDL number MFCD22543729
  • Cas Number : 247135-44-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/5-bromo-4-methyl-2-methylthio-pyridine.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/24566.pdf
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart