AOB640095502 AOBChem

1-[[2-bromo-5-(trifluoromethyl)phenyl]methyl]-piperidine

  • MDL number MFCD22628223
  • Cas Number : 913296-73-8
  • Molecular Weight : 322.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Liquid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/1-2-bromo-5-trifluoromethyl-phenyl-methyl-piperidine.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/24836.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart