AOB640095559 AOBChem

1-(3-Bromo-4-methylbenzyl)pyrrolidine

  • MDL number MFCD22628242
  • Cas Number : 1414870-50-0
  • Molecular Weight : 254.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/1-3-bromo-4-methylbenzyl-pyrrolidine.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/24952.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart