AOB640095747 AOBChem

2,6-Difluoro-3-(methylthio)benzaldehyde

  • MDL number MFCD23713120
  • Cas Number : 1428234-72-3
  • Molecular Weight : 188.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/2-6-difluoro-3-methylthio-benzaldehyde.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/25496.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart