AOB640095837 AOBChem

2-Isoproxy-3-methylpyridine-5-boronic acid pinacol ester

  • MDL number MFCD16996660
  • Cas Number : 1375303-04-0
  • Molecular Weight : 277.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/2-isoproxy-3-methylpyridine-5-boronic-acid-pinacol-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/25757.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart