AOB640095863 AOBChem

5-(3-Fluorophenyl)-1H-tetrazole

  • MDL number MFCD01075702
  • Cas Number : 50907-20-5
  • Molecular Weight : 166.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/5-3-fluorophenyl-1h-tetrazole.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/25808.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart