AOB640095975 AOBChem

N-tert-butyl-2,6-dichloropyridine-4-carboxamide

  • MDL number MFCD11106577
  • Cas Number : 1152504-12-5
  • Molecular Weight : 247.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/n-tert-butyl-2-6-dichloropyridine-4-carboxamide.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/26026.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart