AOB640096123 AOBChem

3-Nitro-4-(trifluoromethoxy)aniline

  • MDL number MFCD03840207
  • Cas Number : 2822-50-6
  • Molecular Weight : 222.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/3-nitro-4-trifluoromethoxy-aniline.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/26361.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart