AOB640096149 AOBChem

4-Bromo-3-formylpyridine HCl

  • MDL number MFCD27756658
  • Cas Number : 1809157-95-6
  • Molecular Weight : 222.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/4-bromo-3-formylpyridine-hcl.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/26427.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart