AOB640096153 AOBChem

4-Bromo-3-pyridinemethanol hydrochloride

  • MDL number MFCD22566376
  • Cas Number : 1546624-18-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/4-bromo-3-pyridinemethanol-hydrochloride.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/26435.pdf
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart