AOB640096228 AOBChem

2,6-difluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-benzenemethanol

  • MDL number MFCD24039809
  • Cas Number : 1417736-43-6
  • Molecular Weight : 270.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/2-6-difluoro-4-4-4-5-5-tetramethyl-1-3-2-dioxaborolan-2-yl-benzenemethanol.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/26596.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart