AOB640096478 AOBChem

3-(Methylthio)-4-(trifluoromethyl)phenylboronic acid pinacol ester

  • MDL number MFCD28015445
  • Cas Number : 2121514-10-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/3-methylthio-4-trifluoromethyl-phenylboronic-acid-pinacol-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/27290.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart