AOB640096512 AOBChem

2-Fluoro-3-(morpholinomethyl)phenylboronic acid

  • MDL number MFCD28142590
  • Cas Number : 2121514-18-7
  • Molecular Weight : 239.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/2-fluoro-3-morpholinomethyl-phenylboronic-acid.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/27365.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart