AOB640096524 AOBChem

3-Bromo-5-methyl-2-(methylthio)pyridine

  • MDL number MFCD22666570
  • Cas Number : 1289049-50-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97.00
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/3-bromo-5-methyl-2-methylthio-pyridine.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/27400.pdf
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart