AOB640096585 AOBChem

2-Chloro-4-(phenoxy)phenylboronic acid

  • MDL number MFCD30344663
  • Cas Number : 2121514-55-2
  • Molecular Weight : 248.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 2-8*C
Physical Form Solid
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/2-chloro-4-phenoxy-phenylboronic-acid.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/27699.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart