AOB640097068 AOBChem

2-Benzyloxy-4,5-dimethylphenylboronic acid, pinacol ester

  • MDL number MFCD29036601
  • Cas Number : 2121514-58-5
  • Molecular Weight : 338.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/2-benzyloxy-4-5-dimethylphenylboronic-acid-pinacol-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/29687.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart