AOB640097089 AOBChem

3-Methyl-1,3-dihydro-2,1-benzoxaborol-1-ol

  • MDL number MFCD27977126
  • Cas Number : 92203-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/3-methyl-1-3-dihydro-2-1-benzoxaborol-1-ol.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/29808.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart