AOB640097136 AOBChem

Methyl 2-chloro-6-(trifluoromethyl)isonicotinate

  • MDL number MFCD16611605
  • Cas Number : 1227594-40-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/methyl-2-chloro-6-trifluoromethyl-isonicotinate.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/30034.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart