AOB640097188 AOBChem

(5-Fluoro-2-isopropoxypyridin-4-yl)methanol

  • MDL number MFCD29088141
  • Cas Number : 1826110-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin China
Product Link https://aobchem.com/5-fluoro-2-isopropoxypyridin-4-yl-methanol.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/30222.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart