AOB640097556 AOBChem

2-Bromo-6-chlorophenylboronic acid pinacol ester

  • MDL number MFCD29092541
  • Cas Number : 2121512-35-2
  • Molecular Weight : 317.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. 2-8*C
Shipping Temperature Room Temperature
Country of Origin CN
Product Link https://aobchem.com/2-bromo-6-chlorophenylboronic-acid-pinacol-ester.html

Safety Information

signalword Warning
pcodes P261 P305 P351 P338
MSDS https://aobchem.com/pub/media/files/sds/31107.pdf
pictograms GHS07
hstatements H315 H319 H335
Availability details for
Quantity
  • Availability
  • Add to Cart